Tisk…
Gelite™ Safe DNA Gel Stain, 10,000X DMSO Solution
Dodavatel: AAT BIOQUEST
17700 17702 17701 17703
AATB17700EA 2940 CZK
AATB17700 AATB17702 AATB17701 AATB17703
Gelite™ Safe DNA Gel Stain, 10,000X DMSO Solution
Barviva Barviva pro elektroforézu
AAT Bioquest is committed to designing our products to be environment-friendly.
  • Improved safety
  • Noncytotoxic
  • High sensitivity
  • Exceptional DNA affinity
  • Robust protocol
  • Multiple applications
Upozornení: For research use only.