Tisk…
2-Aminoethoxypropargyl ddATP (1 µmole)
Dodavatel: AAT BIOQUEST
17084
AATB17084EA 37620 CZK
AATB17084
2-Aminoethoxypropargyl ddATP (1 µmole)
Činidla pro nukleové kyseliny Činidla Sekvenační
Sanger method is one of the most reliable and earliest DNA sequencing methods.
  • In-house quality control
  • Useful for sanger sequencing applications
Upozornení: For research use only.