Tisk…

Aminoallyl UTP sodium salt, (AA-UTP) 4 mM in Tris/EDTA (TE) buffer

Dodavatel: AAT BIOQUEST
Varování

17021
AATB17021EA 12280 CZK
AATB17021
Aminoallyl UTP sodium salt, (AA-UTP) 4 mM in Tris/EDTA (TE) buffer
Aminoallyl UTP sodium salt, (AA-UTP)

The amine-modified uridine 5'-triphosphates (such as aminoallyl-UTP) can be used to produce amine-containing RNA by conventional enzymatic incorporation methods such as transcription, nick translation, random primed labeling, or PCR.

MW: 605.16 g/mol
CAS číslo: 75221-88-4
ADR: 5 II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR