Tisk…
Buccutite™ Rapid APC-iFluor® 750 Tandem Antibody Labelling Kit
Dodavatel: AAT BIOQUEST
1348
AATB1348EA 12380 CZK
AATB1348
Buccutite™ Rapid APC-iFluor® 750 Tandem Antibody Labelling Kit
Systémy pro spojování a značení proteinů
Microscale optimised for labelling 25 µg antibody per reaction. APC-iFluor® 750 tandem is an excellent replacement for APC-Cy7 and APC-Alexa Fluor® 750 tandem since they have almost identical spectra.
  • Robust protocol
  • Consistent results
  • Six-month shelf life
Designed to provide high quality material for downstream fluorescent applications, including flow cytometry.
Informace o dodávce: Shipped with blue ice.
Upozornení: Research use only.