Tisk…
HIS Lite™ iFluor® 568 Tris NTA Chelator
Dodavatel: AAT BIOQUEST
12657
AATB12657EA 16520 CZK
AATB12657
HIS Lite™ iFluor® 568 Tris NTA Chelator
Systémy pro spojování a značení proteinů
iFluor® 568 Tris-NTA Chelator is used as a sensitive fluorescent probe for detecting polyhistidine-labelled proteins in combination with the addition of a heavy metal ion (such as Ni²⁺) in cells, solution and solid surfaces.
  • Stable for 24 months
Quality tested in-house.
Upozornení: For research use only.