Tisk…

D-Luciferin-6-O-β-D-galactopyranoside ≥95%

Dodavatel: AAT BIOQUEST
Varování

12513
AATB12513EA 8190 CZK
AATB12513
D-Luciferin-6-O-β-D-galactopyranoside ≥95%
D-Luciferin-6-O-β-D-galactopyranoside

D-Luciferin galactoside contains a beta-galactoside moiety attached at the 6-O-position, thus preventing it be recognized by the firefly luciferase enzyme.

Vzorec: C₂₂H₂₄O₇
MW: 400.427 g/mol
MDL Number: MFCD09038345
CAS číslo: 131474-38-9
ADR: 5 II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR