Tisk…

iFluor® 555, Maleimide

Dodavatel: AAT BIOQUEST
Varování

1063
AATB1063EA 8190 CZK
AATB1063
iFluor® 555, Maleimide
iFluor®

AAT Bioquest's iFluor® dyes are optimized for labeling proteins, in particular, antibodies.


  • Bright and photostable
  • Minimal quenching
  • Stable conjugation
  • pH independent fluorescence between 3 to 11

ADR: 5,II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR