Tisk…
Amplite® Fluorimetric α-Ketoglutarate Quantitation Kit
Dodavatel: AAT BIOQUEST
10087
AATB10087EA 16380 CZK
AATB10087
Amplite® Fluorimetric α-Ketoglutarate Quantitation Kit
Test Enzymatická stanovení
Alpha-ketoglutarate (α-ketoglutarate) is a key molecule in the Krebs cycle determining the overall rate of the citric acid cycle of the organism.
  • In-house QC testing
  • Sensitive
  • Suggested protocol available
Upozornení: For research use only.