Tisk…

Diethylbenzene (mixture of isomers)

Dodavatel: Thermo Fisher Scientific
Varování

A19548.AE A19548.AP A19548.0F
A19548.AEEA 762 CZK
A19548.AE A19548.AP A19548.0F
Diethylbenzene (mixture of isomers)
Diethylbenzene (mixture of isomers)

CAS No.: 25340-17-4 Molecular Formula: C10H14 Formula Weight: 134.22 Flash Point: 55°(131°F) Refractive Index: 1.495 Boiling Point: 182-184° UN No.: 2049 MDL No.: MFCD00792900

Vzorec: C₁₀H₁₄
MW: 134.22 g/mol
Bod varu: 182…184 °C
Hustota: 0.87
Bod vzplanutí: 55 °C (131 °F)
Storage Temperature: Ambient
MDL Number: MFCD00792900
CAS číslo: 25340-17-4
EINECS: 246-874-9
UN: 2049
ADR: 3 III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR