Tisk…

3-Hydroxy-2-naphthoic acid ≥98%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

Synonyms: 3-Hydroxy-naphthalene-2-carboxylic acid, 2-Hydroxy-3-naphthoic acid

A19214.0B A19214.30
A19214.0BEA 3470 CZK
A19214.0B A19214.30
3-Hydroxy-2-naphthoic acid ≥98%
3-Hydroxy-2-naphthoic acid

Purity: 98% CAS No.: 92-70-6 Molecular Formula: C11H8O3 Formula Weight: 188.18 Flash Point: 220°(428°F) Melting Point: 218-222° Beilstein Reference: 744100 Merck Reference: 14,4835 MDL No.: MFCD00004103

Vzorec: C₁₁H₈O₃
MW: 188.18
Bod varu: 368
Bod tání: 218…222 °C
Bod vzplanutí: 220 °C (428 °F)
Storage Temperature: Ambient
MDL Number: MFCD00004103
CAS číslo: 92-70-6
EINECS: 202-180-8
Merck Index: 13,04860

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR