Tisk…

(±)-3-Methylvaleric acid ≥99%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

Synonyms: 3-Methylvaleric Acid, 3-Methylpentanoic acid, (±)-3-Methylpentanoic acid

A18886.06 A18886.14 A18886.22
A18886.06EA 1170 CZK
A18886.06 A18886.14 A18886.22
(±)-3-Methylvaleric acid ≥99%
(±)-3-Methylvaleric acid

Purity: 99% CAS No.: 105-43-1 Molecular Formula: C6H12O2 Formula Weight: 116.16 Flash Point: 85°(185°F) Refractive Index: 1.4160 Odour: Unpleasant Melting Point: -42° Boiling Point: 196-198° Beilstein Reference: 1720696 UN No.: 3265 MDL No.: MFCD00002729

Vzorec: C₆H₁₂O₂
MW: 116.16
Bod varu: 196…198 °C
Bod tání: –42 °C
Hustota: 0.93
Bod vzplanutí: 85 °C (185 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002729
CAS číslo: 105-43-1
EINECS: 203-297-7
UN: 3265

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR