Tisk…

4-Benzoyl-4'-bromobiphenyl ≥99%

Dodavatel: Thermo Fisher Scientific

A18482.14 A18482.22
A18482.14EA 1890 CZK
A18482.14 A18482.22
4-Benzoyl-4'-bromobiphenyl ≥99%
4-Benzoyl-4'-bromobiphenyl

Purity: 99% CAS No.: 63242-14-8 Molecular Formula: C19H13BrO Formula Weight: 337.22 Melting Point: 155-158° Beilstein Reference: 2378429

Vzorec: C₁₉H₁₃BrO
Bod tání: 155…158 °C
Storage Temperature: Ambient
MDL Number: MFCD00209630
CAS číslo: 63242-14-8

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR