Tisk…

4,4'-Bis(dimethylamino)diphenylmethane ≥98%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

Synonyms: 4,4'-Methylenebis(N,N-dimethylaniline), Bis[4-(dimethylamino)phenyl]methane

A18466.18 A18466.30 A18466.09
A18466.18EA 1580 CZK
A18466.18 A18466.30 A18466.09
4,4'-Bis(dimethylamino)diphenylmethane ≥98%
4,4'-Bis(dimethylamino)diphenylmethane

Purity: 98+% CAS No.: 101-61-1 Molecular Formula: C17H22N2 Formula Weight: 254.38 Flash Point: 178°(352°F) Melting Point: 85-88° Boiling Point: 155-157°/0.1mm Beilstein Reference: 479956 Merck Reference: 14,6176 UN No.: 3077 MDL No.: MFCD00008317

Vzorec: C₁₇H₂₂N₂
Bod varu: 155…157 °C (0,1 mmHg)
Bod tání: 85…88 °C
Bod vzplanutí: 178 °C (352 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008317
CAS číslo: 101-61-1
EINECS: 202-959-2
ADR: 9,III
Merck Index: 14,6176

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR