Tisk…

Stearic acid hydrazide ≥97%

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: Stearicacidhydrazide

A18291.09 A18291.18 A18291.30
A18291.09EA 1350 CZK
A18291.09 A18291.18 A18291.30
Stearic acid hydrazide ≥97%
Stearic acid hydrazide

Purity: 97% CAS No.: 4130-54-5 Molecular Formula: CH3(CH2)16CONHNH2 Formula Weight: 298.51 Melting Point: 114-116° MDL No.: MFCD00014764

Vzorec: C₁₈H₃₈N₂O
MW: 298.51 g/mol
Bod tání: 114…116 °C
Storage Temperature: Ambient
MDL Number: MFCD00014764
CAS číslo: 4130-54-5
EINECS: 223-946-8

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR