Tisk…

4,4'-Bi-o-toluidin dihydrochlorid ≥97%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

A18196.14 A18196.22
A18196.14EA 1830 CZK
A18196.14 A18196.22
4,4'-Bi-o-toluidin dihydrochlorid ≥97%
4,4'-Bi-o-toluidin dihydrochlorid

Purity: 97% CAS No.: 612-82-8 Molecular Formula: C14H16N2.2HCl Formula Weight: 285.22 Odour: Odourless Melting Point: 300° UN No.: 3077 MDL No.: MFCD00012960

Vzorec: C₁₄H₁₈Cl₂N₂
Bod tání: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00012960
CAS číslo: 612-82-8
EINECS: 210-322-5
ADR: 9,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR