Tisk…

2-Amino-2-methylpropanol, (max. 5% H₂O) 95%

Dodavatel: Thermo Fisher Scientific
Varování

A17814.0F A17814.AE A17814.AP
A17814.0FEA 4470 CZK
A17814.0F A17814.AE A17814.AP
2-Amino-2-methylpropanol, (max. 5% H₂O) 95%
2-Amino-2-methylpropanol

Purity: 95% CAS No.: 124-68-5 Molecular Formula: C4H11NO Formula Weight: 89.14 Flash Point: 67°(152°F) Refractive Index: 1.4455 Odour: Amine-like pH: 11.3 Melting Point: 24-28° Boiling Point: 164-166° Beilstein Reference: 505979 Merck Reference: 14,449 MDL No.: MFCD00008051

Vzorec: (CH₃)₂C(NH₂)CH₂OH
Bod varu: 164…166 °C
Bod tání: 24…28 °C
Hustota: 0,934
Bod vzplanutí: 67 °C (152 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008051
CAS číslo: 124-68-5
EINECS: 204-709-8
Merck Index: 14,449

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR