Tisk…

DL-Glutamic acid monohydrate ≥99%

Dodavatel: Thermo Fisher Scientific

Synonyms: DL-Glutamic Acid Monohydrate

A17719.14 A17719.22 A17719.36
A17719.14EA 1010 CZK
A17719.14 A17719.22 A17719.36
DL-Glutamic acid monohydrate ≥99%
DL-Glutamic acid monohydrate

Purity: 99% CAS No.: 19285-83-7 Molecular Formula: C5H9NO4.H2O Formula Weight: 165.15 (147.13anhy) Melting Point: ca 200° dec. Beilstein Reference: 4155097 Merck Reference: 14,4469 MDL No.: MFCD00150703

Vzorec: C₅H₉NO₄·1H₂O
MW: 165.15 g/mol (147.13 anhy)
Bod tání: ∼ 200 °C (rozkl.)
Hustota: 1.460
Storage Temperature: Ambient
MDL Number: MFCD00150703
CAS číslo: 19285-83-7
EINECS: 210-522-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR