Tisk…

1-Naphthyl acetate ≥99%

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: Naphthalen-1-yl acetate, Ethanoic acid 1-naphthyl ester, Acetic acid 1-naphthyl ester, α-Naphthyl acetate, Acetic acid α-naphthyl ester, Ethanoic acid α-naphthyl ester

A17482.18 A17482.30
A17482.18EA 1850 CZK
A17482.18 A17482.30
1-Naphthyl acetate ≥99%
1-Naphthyl acetate

Purity: 99% CAS No.: 830-81-9 Molecular Formula: C12H10O2 Formula Weight: 186.21 Storage Temperature: 2°C to 8°C Flash Point: 110°(230°F) Melting Point: 43-46° Beilstein Reference: 2046403 MDL No.: MFCD00003922

Vzorec: C₁₂H₁₀O₂
Bod varu: 310 °C (1013 hPa)
Bod tání: 43…46 °C
Hustota: 1,1305 g/cm³ (50 °C)
Bod vzplanutí: >110 °C (230 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00003922
CAS číslo: 830-81-9
EINECS: 212-599-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR