Tisk…

N-2-Aminoethyl-1-naftylamin dihydrochlorid 96%

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: N-2-Aminoethyl-1-naftylamin dihydrochlorid

A17164.09 A17164.18
A17164.09EA 1880 CZK
A17164.09 A17164.18
N-2-Aminoethyl-1-naftylamin dihydrochlorid 96%
N-(1-Naphthyl)ethylenediamine dihydrochlorid

Purity: 96% CAS No.: 1465-25-4 Molecular Formula: C12H14N2.2HCl Formula Weight: 259.18 Melting Point: 191-199° Beilstein Reference: 3707471 Merck Reference: 14,6409 MDL No.: MFCD00012556

Vzorec: C₁₂H₁₆Cl₂N₂
Bod tání: 191…199 °C
Storage Temperature: Ambient
MDL Number: MFCD00012556
CAS číslo: 1465-25-4
EINECS: 215-981-2
Merck Index: 14,6409
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3643e6df3e9cf8dc627823e9090c97004ee25308&vid=a30fc28279c11af12c6b735b00b84fc8f08caf34&prid=web&clid=SAPDAM

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR