Tisk…

3-Bromo-5-iodobenzoic acid ≥97%

Dodavatel: Thermo Fisher Scientific
Varování

A17076.14 A17076.06 A17076.22
A17076.14EA 6560 CZK
A17076.14 A17076.06 A17076.22
3-Bromo-5-iodobenzoic acid ≥97%
3-Bromo-5-iodobenzoic acid

Purity: 97% CAS No.: 188815-32-9 Molecular Formula: C7H4BrIO2 Formula Weight: 326.92 Melting Point: 218-221° Beilstein Reference: 3240837

Vzorec: C₇H₄BrIO₂
MW: 326.92
Bod tání: 218…221 °C
Storage Temperature: Ambient
MDL Number: MFCD00191851
CAS číslo: 188815-32-9

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR