Tisk…

Adenine sulphate ≥98%

Dodavatel: Thermo Fisher Scientific
Varování

A16964.09 A16964.18
A16964.09EA 1370 CZK
A16964.09 A16964.18
Adenine sulphate ≥98%
Adenine sulphate

Purity: 98+% CAS No.: 321-30-2 Molecular Formula: C10H10N10.H2SO4 Formula Weight: 368.34 Odour: Odourless Melting Point: ca 210° dec. Beilstein Reference: 3820263 Merck Reference: 14,152 MDL No.: MFCD00213655

Vzorec: C₅H₅N₅·½H₂SO₄
Storage Temperature: Ambient
MDL Number: MFCD00213655
CAS číslo: 321-30-2
EINECS: 206-286-5
Merck Index: 14,152

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR