Tisk…

Benzoan lithný 99

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: Benzoic acid lithium salt

A16571.30 A16571.0B
A16571.30EA 3850 CZK
A16571.30 A16571.0B
Benzoan lithný 99
Benzoan lithný

Purity: 99+% CAS No.: 553-54-8 Molecular Formula: C7H5LiO2 Formula Weight: 128.06 Odour: Odourless Melting Point: 300° Beilstein Reference: 3598089 Merck Reference: 14,5523 MDL No.: MFCD00035540

Vzorec: C₇H₅LiO₂
MW: 128.06
Bod tání: >300 °C
Hustota: 1.397 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00035540
CAS číslo: 553-54-8
EINECS: 209-042-6
Merck Index: 12,05547

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR