Tisk…

(-)-Dibenzoyl-L-tartaric acid monohydrate ≥98%

Dodavatel: Thermo Fisher Scientific
Varování

A16180.0E A16180.22 A16180.36
A16180.0EEA 13490 CZK
A16180.0E A16180.22 A16180.36
(-)-Dibenzoyl-L-tartaric acid monohydrate ≥98%
(-)-Dibenzoyl-L-tartaric acid monohydrate

Purity: 98+% CAS No.: 62708-56-9 Molecular Formula: C18H14O8.H2O Formula Weight: 376.33 (358.31anhy) Melting Point: 90-92° MDL No.: MFCD00149119

Vzorec: C₁₈H₁₄O₈·1H₂O
Bod tání: 90…92 °C
Storage Temperature: Ambient
MDL Number: MFCD00149119
CAS číslo: 62708-56-9
EINECS: 613-072-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR