Tisk…

EGTA (ethylene glycol bis(2-aminoethyl ether)-N,N,N',N'-tetraacetic acid) 97%

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: EGTA, Ethylenebis(oxyethylenenitrilo)tetra(acetic acid), Kestranal® FE PA, Idranal® VI, Ethylene glycol-O,O'-bis(2-aminoethyl)-N,N,N',N'-tetraacetic acid, Ethylenebis(oxyethylenenitrilo)tetraacetic acid, Ethylene Glycol Bis(2-aminoethyl Ether)-N,N,N,N-tetraacetic acid, 3,12-Bis(carboxymethyl)-6,9-dioxa-3,12-diazatetradecanedioic acid, Titriplex® VI

A16086.09 A16086.18 A16086.30
A16086.09EA 1730 CZK
A16086.09 A16086.18 A16086.30
EGTA (ethylene glycol bis(2-aminoethyl ether)-N,N,N',N'-tetraacetic acid) 97%
EGTA (ethylene glycol bis(2-aminoethyl ether)-N,N,N',N'-tetraacetic acid)

Purity: 97% CAS No.: 67-42-5 Molecular Formula: C14H24N2O10 Formula Weight: 380.35 Melting Point: ca 242° dec. Beilstein Reference: 1717370 Merck Reference: 14,3529 MDL No.: MFCD00004291

Vzorec: C₁₄H₂₄N₂O₁₀
MW: 380.35
Bod tání: ∼ 242 °C (rozkl.)
Storage Temperature: Ambient
MDL Number: MFCD00004291
CAS číslo: 67-42-5
EINECS: 200-651-2
Merck Index: 13,03560

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR