Tisk…

trans-2-Octen-1-ol ≥97%

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: Trans-2-octen-1-ol, trans-2-Octen-1-ol, trans-Oct-2-en-1-ol

A15960.06 A15960.14 A15960.22
A15960.06EA 1630 CZK
A15960.06 A15960.14 A15960.22
trans-2-Octen-1-ol ≥97%
trans-2-Octen-1-ol

Purity: 97% CAS No.: 18409-17-1 Molecular Formula: C8H16O Formula Weight: 128.22 Flash Point: 85°(185°F) Refractive Index: 1.4465 Boiling Point: 88°/11mm Beilstein Reference: 1720707 MDL No.: MFCD00014057

Vzorec: C₈H₁₆O
Bod varu: 88 °C (11 mmHg)
Hustota: 0,850
Bod vzplanutí: 85 °C (185 °F)
Storage Temperature: Ambient
MDL Number: MFCD00014057
CAS číslo: 18409-17-1
EINECS: 242-288-2
UN: 1987
ADR: 3,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR