Tisk…

2-Merkaptoethanol ≥98%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

A15890.0B A15890.0I A15890.30
A15890.0BEA 3010 CZK
A15890.0B A15890.0I A15890.30
2-Merkaptoethanol ≥98%
2-Merkaptoethanol

Purity: 98+% CAS No.: 60-24-2 Molecular Formula: HSCH2CH2OH Formula Weight: 78.13 Flash Point: 73°(163°F) Refractive Index: 1.500 Odour: Unpleasant Melting Point: -100° Boiling Point: 156-158° Beilstein Reference: 773648 Merck Reference: 14,5869 UN No.: 2966 MDL No.: MFCD00004890

Vzorec: HSCH₂CH₂OH
Bod varu: 156…158 °C
Bod tání: –100 °C
Hustota: 1,114
Bod vzplanutí: 73 °C (163 °F)
Storage Temperature: Ambient
MDL Number: MFCD00004890
CAS číslo: 60-24-2
EINECS: 200-464-6
UN: 2966
ADR: 6.1,II
Merck Index: 14,5869
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cc427209b84d0343a5ae47c435ddfc53df4013d2&vid=7d70fd087d948d54e7a946d3b2310e2d820d2396&prid=web&clid=SAPDAM

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR