Tisk…

Cholinchlorid ≥98%

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: 2-hydroxy-N,N,N-trimethylethanaminium chloride

A15828.22 A15828.0E A15828.36
A15828.22EA 1040 CZK
A15828.22 A15828.0E A15828.36
Cholinchlorid ≥98%
Cholinchlorid

Purity: 98+% CAS No.: 67-48-1 Molecular Formula: HOCH2CH2N(CH3)3Cl Formula Weight: 139.63 Physical Form: Solid Sensitivity: Hygroscopic Beilstein Reference: 3563126 Merck Reference: 14,2206 MDL No.: MFCD00011721


An acyl group acceptor

Vzorec: HOH₂CCH₂N(CH₃)₃Cl
MW: 139.62
Bod tání: ∼ 305 °C (rozkl.)
Hustota: ∼1.205 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011721
CAS číslo: 67-48-1
EINECS: 200-655-4
Merck Index: 13,02226
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=77b015e76f45084bc46da3249578a85652cd6802&vid=85c8a0a1b152b861b3bc3ec8fe25a70509221d85&prid=web&clid=SAPDAM

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR