Tisk…

Sodium chlorodifluoroacetate 97%, rods

Dodavatel: Thermo Fisher Scientific
Varování

A15756.14 A15756.22 A15756.36
A15756.14EA 2170 CZK
A15756.14 A15756.22 A15756.36
Sodium chlorodifluoroacetate 97%, rods
Sodium chlorodifluoroacetate

Purity: 97% CAS No.: 1895-39-2 Molecular Formula: ClCF2CO2Na Formula Weight: 152.46 Melting Point: ca 195° dec. Beilstein Reference: 4162363

Vzorec: C₂ClF₂NaO₂
Storage Temperature: Ambient
MDL Number: MFCD00064771
CAS číslo: 1895-39-2
EINECS: 217-586-0

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR