Tisk…

α-chlor-4-nitrotoluen ≥99%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

A15749.30
A15749.30EA 9270 CZK
A15749.30
α-chlor-4-nitrotoluen ≥99%
α-chlor-4-nitrotoluen

Purity: 99% CAS No.: 100-14-1 Molecular Formula: C7H6ClNO2 Formula Weight: 171.58 Melting Point: 70-74° Beilstein Reference: 387187 UN No.: 3261 MDL No.: MFCD00007374

Vzorec: C₇H₆ClNO₂
Bod varu: 293 °C (1013 hPa)
Bod tání: 70…74 °C
Hustota: 1,5647 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007374
CAS číslo: 100-14-1
EINECS: 202-822-7
UN: 3261
ADR: 8,II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR