Tisk…

Octadecyltrichlorosilane 95% cont. 5-10% branched isomers

Dodavatel: Thermo Fisher Scientific
Nebezpečí

A15732.0B A15732.18 A15732.30
A15732.0BEA 12700 CZK
A15732.0B A15732.18 A15732.30
Octadecyltrichlorosilane 95% cont. 5-10% branched isomers
Octadecyltrichlorosilane

Purity: 95% CAS No.: 112-04-9 Molecular Formula: C18H37Cl3Si Formula Weight: 387.94 Flash Point: 189°(372°F) Refractive Index: 1.4600 Odour: Pungent Melting Point: ca 20° Boiling Point: 160-162°/3mm Beilstein Reference: 1778847 UN No.: 1800 MDL No.: MFCD00000484

Vzorec: C₁₈H₃₇Cl₃Si
Bod varu: 160…162 °C (3 mmHg)
Bod tání: ∼ 20 °C
Hustota: 0,984
Bod vzplanutí: 189 °C (372 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000484
CAS číslo: 112-04-9
EINECS: 203-930-7
UN: 1800
ADR: 8,II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR