Tisk…

dicyklohexylamin 98%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

Synonyms: DCHA

A15671.0F A15671.AE A15671.AP
A15671.0FEA 3160 CZK
A15671.0F A15671.AE A15671.AP
dicyklohexylamin 98%
dicyklohexylamin

Purity: 98% CAS No.: 101-83-7 Molecular Formula: C12H23N Formula Weight: 181.32 Flash Point: 99°(210°F) Refractive Index: 1.4842 Melting Point: -2° Boiling Point: 256° Beilstein Reference: 605923 Merck Reference: 14,3095 UN No.: 2565 MDL No.: MFCD00011658

Vzorec: C₁₂H₂₃N
Bod varu: 256 °C
Bod tání: –2 °C
Hustota: 0,913
Bod vzplanutí: 99 °C (210 °F)
Storage Temperature: Ambient
MDL Number: MFCD00011658
CAS číslo: 101-83-7
EINECS: 202-980-7
UN: 2565
ADR: 8,III
Merck Index: 14,3095

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR