Tisk…

D-(-)-α-Phenylglycine ≥99%

Dodavatel: Thermo Fisher Scientific

A15669.14 A15669.22 A15669.36
A15669.14EA 920 CZK
A15669.14 A15669.22 A15669.36
D-(-)-α-Phenylglycine ≥99%
D-(-)-α-Phenylglycine

Purity: 99% CAS No.: 875-74-1 Molecular Formula: C8H9NO2 Formula Weight: 151.17 Melting Point: 277° dec. Beilstein Reference: 2208676 Merck Reference: 14,7291 Optical Rotation: -154° (c=1 in 1N HCl) MDL No.: MFCD00008061

Vzorec: C₈H₉NO₂
Storage Temperature: Ambient
MDL Number: MFCD00008061
CAS číslo: 875-74-1
EINECS: 212-876-3
Merck Index: 14,7291

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR