Tisk…

2,2-Dimethyl-1,3-dioxane-4,6-dione (isopropylidene malonate, meldrum's acid) ≥98%

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: 2,2-Dimethyl-1,3-dioxane-4,6-dione

A15603.14 A15603.22 A15603.36 AB117158.0500
A15603.14EA 1540 CZK
A15603.14 A15603.22 A15603.36 ABCRAB117158.0500
2,2-Dimethyl-1,3-dioxane-4,6-dione (isopropylidene malonate, meldrum's acid) ≥98%
2,2-Dimethyl-1,3-dioxane-4,6-dione (isopropylidene malonate, meldrum's acid)

Purity: 98% CAS No.: 2033-24-1 Molecular Formula: C6H8O4 Formula Weight: 144.13 Melting Point: 92-96° Beilstein Reference: 117310 Merck Reference: 14,5817 MDL No.: MFCD00006638

Vzorec: C₆H₈O₄
Bod tání: 92…96 °C
Storage Temperature: Ambient
MDL Number: MFCD00006638
CAS číslo: 2033-24-1
EINECS: 217-992-8
Merck Index: 14,5817

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR