Tisk…

Aminoacetaldehyde dimethyl acetal 99%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

A15498.14 A15498.22 A15498.36
A15498.14EA 1450 CZK
A15498.14 A15498.22 A15498.36
Aminoacetaldehyde dimethyl acetal 99%
Aminoacetaldehyde dimethyl acetal

Purity: 99% CAS No.: 22483-09-6 Molecular Formula: H2NCH2CH(OCH3)2 Formula Weight: 105.14 Flash Point: 41°(106°F) Refractive Index: 1.4170 Boiling Point: 135-138° Beilstein Reference: 741868 UN No.: 2733 MDL No.: MFCD00008135

Vzorec: C₄H₁₁NO₂
Bod varu: 135…138 °C
Hustota: 0,973
Bod vzplanutí: 41 °C (106 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008135
CAS číslo: 22483-09-6
EINECS: 245-026-5
ADR: 3,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR