Tisk…

3-Pentanon ≥99%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

A15297.0F A15297.AE A15297.AP
A15297.0FEA 3640 CZK
A15297.0F A15297.AE A15297.AP
3-Pentanon ≥99%
3-Pentanone

Purity: 99% CAS No.: 96-22-0 Molecular Formula: C5H10O Formula Weight: 86.13 Flash Point: 7°(45°F) Refractive Index: 1.3920 Odour: Acetone-like Melting Point: -39° Boiling Point: 102-103° Beilstein Reference: 635749 Merck Reference: 14,3121 UN No.: 1156 MDL No.: MFCD00009320

Vzorec: C₅H₁₀O
Bod varu: 102…103 °C
Bod tání: –39 °C
Hustota: 0,812
Bod vzplanutí: 7 °C (45 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009320
CAS číslo: 96-22-0
EINECS: 202-490-3
UN: 1156
ADR: 3,II
Merck Index: 14,3121

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR