Tisk…

Fluorid boritý - ether komplex ≥98%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

Synonyms: Boron fluoride, ethyl ether complex, Boron trifluoride ethyl etherate, Boron trifluoride diethyl ether, Boron trifluoride etherate, Diethyl ether boron trifluoride, Boron trifluoride diethyl etherate

A15275.AE A15275.AP A15275.AU
A15275.AEEA 1100 CZK
A15275.AE A15275.AP A15275.AU
Fluorid boritý - ether komplex ≥98%
Fluorid boritý - ether komplex

Purity: 98+% CAS No.: 109-63-7 Molecular Formula: C4H10BF3O Formula Weight: 141.93 Flash Point: 47°(116°F) Refractive Index: 1.3440 Odour: Acrid Melting Point: -60° Boiling Point: 125-126° Beilstein Reference: 3909607 Merck Reference: 14,1350 UN No.: 2604 MDL No.: MFCD00013194

Vzorec: C₄H₁₀BF₃O
Bod varu: 125…126 °C
Bod tání: –60 °C
Hustota: 1,130
Bod vzplanutí: 47 °C (116 °F)
Storage Temperature: Ambient
MDL Number: MFCD00013194
CAS číslo: 109-63-7
EINECS: 203-689-8
UN: 2604
ADR: 8,I
Merck Index: 14,1350

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR