Tisk…

Thiophene-2-carboxamide ≥99%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

A15219.06 A15219.14
A15219.06EA 1740 CZK
A15219.06 A15219.14
Thiophene-2-carboxamide ≥99%
Thiophene-2-carboxamide

Purity: 99% CAS No.: 5813-89-8 Molecular Formula: C5H5NOS Formula Weight: 127.17 Melting Point: 178-183° Beilstein Reference: 110153

Vzorec: C₅H₅NOS
Bod tání: 178…183 °C
Storage Temperature: Ambient
MDL Number: MFCD00010425
CAS číslo: 5813-89-8

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR