Tisk…

5-Iodoisatin ≥97%

Dodavatel: Thermo Fisher Scientific

Synonyms: 5-Iodoindoline-2,3-dione, 5-Iodo-1H-indole-2,3-dione

A14988.06
A14988.06EA 1720 CZK
A14988.06
5-Iodoisatin ≥97%
5-Iodoisatin

Purity: 97% CAS No.: 20780-76-1 Molecular Formula: C8H4INO2 Formula Weight: 273.03 Melting Point: ca 276° dec. Beilstein Reference: 131248

Vzorec: C₈H₄INO₂
MW: 273.03
Bod tání: ∼ 276 °C (rozkl.)
Storage Temperature: Ambient
MDL Number: MFCD00016899
CAS číslo: 20780-76-1
EINECS: 244-035-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR