Tisk…

Adenin ≥99%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

Synonyms: Purin-6-amine, 7H-Purin-6-amine, 6-Aminopurine

A14906.14 A14906.18 A14906.30 A14906.36 A14906.A7
A14906.14EA 1960 CZK
A14906.14 A14906.18 A14906.30 A14906.36 A14906.A7
Adenin ≥99%
Adenin

Purity: 99% CAS No.: 73-24-5 Molecular Formula: C5H5N5 Formula Weight: 135.13 Odour: Odourless Melting Point: 360° dec. Beilstein Reference: 5777 Merck Reference: 14,152 UN No.: 2811 MDL No.: MFCD00041790

Vzorec: C₅H₅N₅
MW: 135.13
Bod tání: >360 °C (rozkl.)
Hustota: 0.99741 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00041790
CAS číslo: 73-24-5
EINECS: 200-796-1
UN: 2811
ADR: 6.1 III
Merck Index: 13,00152

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR