Tisk…

2-(Trifluoromethyl)phenylhydrazine ≥97%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

Synonyms: 1-(2-(Trifluoromethyl)phenyl)hydrazine, 2-Hydrazinobenzotrifluoride

A14781.03 A14781.06
A14781.03EA 1320 CZK
A14781.03 A14781.06
2-(Trifluoromethyl)phenylhydrazine ≥97%
2-(Trifluoromethyl)phenylhydrazine

Purity: 97% CAS No.: 365-34-4 Molecular Formula: C7H7F3N2 Formula Weight: 176.14 Melting Point: 57-63° Beilstein Reference: 641592

Vzorec: C₇H₇F₃N₂
MW: 176.14
Bod tání: 57…63 °C
Storage Temperature: Ambient
MDL Number: MFCD00052687
CAS číslo: 365-34-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR