Tisk…

Kyselina 6-aminokaprová 99%

Dodavatel: Thermo Fisher Scientific
Varování

A14719.0E A14719.22 A14719.36
A14719.0EEA 13320 CZK
A14719.0E A14719.22 A14719.36
Kyselina 6-aminokaprová 99%
Kyselina 6-aminokaprová

Purity: 99% CAS No.: 60-32-2 Molecular Formula: C6H13NO2 Formula Weight: 131.18 Physical Form: Cryst powder Appearance: White Melting Point: 210-212° Beilstein Reference: 906872 Merck Reference: 14,432 MDL No.: MFCD00008238

Vzorec: C₆H₁₃NO₂
Bod varu: 257 °C (1013 hPa)
Bod tání: 210…212 °C
Bod vzplanutí: 207 °C
Storage Temperature: Ambient
MDL Number: MFCD00008238
CAS číslo: 60-32-2
EINECS: 200-469-3
Merck Index: 14,432

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR