Tisk…

4-Decyloxybenzoic acid 96%

Dodavatel: Thermo Fisher Scientific

A14703.06 A14703.14 A14703.22
A14703.06EA 1220 CZK
A14703.06 A14703.14 A14703.22
4-Decyloxybenzoic acid 96%
4-Decyloxybenzoic acid

Purity: 96% CAS No.: 5519-23-3 Molecular Formula: C17H26O3 Formula Weight: 278.39 Melting Point: 97°(S) Beilstein Reference: 2113730

Vzorec: C₁₇H₂₆O₃
Bod tání: 97 °C (subl.)
Storage Temperature: Ambient
MDL Number: MFCD00020360
CAS číslo: 5519-23-3

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR