Tisk…

Crocein scarlet 3B

Dodavatel: Thermo Fisher Scientific

A14642.18 A14642.30
A14642.18EA 1180 CZK
A14642.18 A14642.30
Crocein scarlet 3B
Crocein scarlet 3B

CAS No.: 5413-75-2 Molecular Formula: C22H14N4Na2O7S2 Formula Weight: 556.49 Solubility: Soluble in water MDL No.: MFCD00003907

Vzorec: C₂₂H₁₆N₄O₇S₂
Storage Temperature: Ambient
MDL Number: MFCD00003907
CAS číslo: 5413-75-2
EINECS: 226-502-1

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR