Tisk…

4,4'-Dimethoxybenzhydrol ≥98%

Dodavatel: Thermo Fisher Scientific

A14604.09 A14604.18
A14604.09EA 1050 CZK
A14604.09 A14604.18
4,4'-Dimethoxybenzhydrol ≥98%
4,4'-Dimethoxybenzhydrol

Purity: 98+% CAS No.: 728-87-0 Molecular Formula: C15H16O3 Formula Weight: 244.29 Melting Point: 69-71° Beilstein Reference: 748532

Vzorec: C₁₅H₁₆O₃
Bod varu: 362 °C (1013 hPa)
Bod tání: 69…71 °C
Storage Temperature: Ambient
MDL Number: MFCD00008410
CAS číslo: 728-87-0
EINECS: 211-975-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR