Tisk…

Sulfid kademnatý ≥98%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

A14544.22 A14544.0E A14544.36
A14544.22EA 989 CZK
A14544.22 A14544.0E A14544.36
Sulfid kademnatý ≥98%
Sulfid kademnatý

Purity: 98% CAS No.: 1306-23-6 Molecular Formula: CdS Formula Weight: 144.46 Odour: Odourless Melting Point: 980° subl. UN No.: 3077 MDL No.: MFCD00010922

Vzorec: CdS
Bod tání: 980 °C (subl.)
Hustota: 4,82
Storage Temperature: Ambient
MDL Number: MFCD00010922
CAS číslo: 1306-23-6
EINECS: 215-147-8
UN: 3077
ADR: 9,III
Merck Index: 14,1628

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR