Tisk…

2-Bromobenzoyl chloride ≥98%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

Synonyms: 2-Bromobenzene-1-carbonyl chloride, o-Bromobenzoyl chloride

A14520.09 A14520.18 A14520.30
A14520.09EA 1230 CZK
A14520.09 A14520.18 A14520.30
2-Bromobenzoyl chloride ≥98%
2-Bromobenzoyl chloride

Purity: 98% CAS No.: 7154-66-7 Molecular Formula: C7H4BrClO Formula Weight: 219.47 Flash Point: 107°(224°F) Refractive Index: 1.5960 Melting Point: 10° Boiling Point: 245° Beilstein Reference: 508506 UN No.: 3265 MDL No.: MFCD00000655

Vzorec: C₇H₄BrClO
MW: 219.46
Bod varu: 245 °C
Bod tání: 10 °C
Hustota: 1.674
Bod vzplanutí: 107 °C (224 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000655
CAS číslo: 7154-66-7
EINECS: 230-507-4
UN: 3265
ADR: 8 II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR