Tisk…

1-Jódpropan ≥98% stabilizovaný

Dodavatel: Thermo Fisher Scientific
Varování


Stabilization: Stabilised with copper

A14388.22 A14388.36
A14388.22EA 1460 CZK
A14388.22 A14388.36
1-Jódpropan ≥98% stabilizovaný
1-Jódpropan

Stabilized with copper


Purity: 98+% CAS No.: 107-08-4 Molecular Formula: C3H7I Formula Weight: 169.99 Flash Point: 44°(111°F) Refractive Index: 1.5050 Melting Point: -101° Boiling Point: 101-102° Beilstein Reference: 505937 Merck Reference: 14,7862 UN No.: 2392 MDL No.: MFCD00001095

Vzorec: C₃H₇I
Bod varu: 101…102 °C
Bod tání: –101 °C
Hustota: 1,744
Bod vzplanutí: 44 °C (111 °F)
Storage Temperature: Ambient
MDL Number: MFCD00001095
CAS číslo: 107-08-4
EINECS: 203-460-2
UN: 2392
ADR: 3,III
Merck Index: 14,7862

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR