Tisk…

Fenylacetaldehyd 95%

Dodavatel: Thermo Fisher Scientific
Varování

A14263.14 A14263.22 A14263.36
A14263.14EA 872 CZK
A14263.14 A14263.22 A14263.36
Fenylacetaldehyd 95%
Fenylacetaldehyd

Purity: 95% CAS No.: 122-78-1 Molecular Formula: C8H8O Formula Weight: 120.15 Storage Temperature: 2°C to 8°C Flash Point: 68°(154°F) Refractive Index: 1.5350 Boiling Point: 194-196° Beilstein Reference: 385791 Merck Reference: 14,7265 MDL No.: MFCD00006993

Vzorec: C₈H₈O
Bod varu: 194…196 °C
Bod tání: 33…34 °C
Hustota: 1,075
Bod vzplanutí: 68 °C (154 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00006993
CAS číslo: 122-78-1
EINECS: 204-574-5
Merck Index: 14,7265

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR