Tisk…

L(+)-Glutamin 99%

Dodavatel: Thermo Fisher Scientific

A14201.0B A14201.18 A14201.30 AB116930.0050
A14201.0BEA 11500 CZK
A14201.0B A14201.18 A14201.30 ABCRAB116930.0050
L(+)-Glutamin 99%
L(+)-Glutamin

Purity: 99% CAS No.: 56-85-9 Molecular Formula: C5H10N2O3 Formula Weight: 146.15 Physical Form: Powder Appearance: White Melting Point: 185° dec. Beilstein Reference: 1723797 Merck Reference: 14,4471 Optical Rotation: +33° (c=5 in 5N HCl) MDL No.: MFCD00008044

Vzorec: C₅H₁₀N₂O₃
Bod varu: 355 °C (1013 hPa)
Hustota: 1,5326 g/cm³ (130 °C)
Storage Temperature: Ambient
MDL Number: MFCD00008044
CAS číslo: 56-85-9
EINECS: 200-292-1
Merck Index: 14,4471
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cb7a812ec5d2c73a797fc464926df9eef12c5c9a&vid=15d7d87daff0fec33a974a578415e157a3278049&prid=web&clid=SAPDAM

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR