Tisk…

N,N-Dimethyl-p-fenylendiamoniumsulfát 98%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

A14175.14 A14175.22 A14175.36
A14175.14EA 2280 CZK
A14175.14 A14175.22 A14175.36
N,N-Dimethyl-p-fenylendiamoniumsulfát 98%
N,N-Dimethyl-p-fenylendiamoniumsulfát

Purity: 98% CAS No.: 536-47-0 Molecular Formula: C8H12N2.H2SO4 Formula Weight: 234.28 Odour: Odourless Melting Point: 200-205° dec. Beilstein Reference: 4612278 UN No.: 2811 MDL No.: MFCD00012992

Vzorec: C₈H₁₄N₂O₄S
Storage Temperature: Ambient
MDL Number: MFCD00012992
CAS číslo: 536-47-0
EINECS: 208-636-2
UN: 2811
ADR: 6.1,II

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR